missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) iodide, ultra dry, 99.99% (metals basis)

Product Code. 11376878
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376878 1 g 1g
11386878 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376878 Supplier Thermo Scientific Alfa Aesar Supplier No. 013619.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Light and Air Sensitive and Hygroscopic | CAS: 7783-86-0 | FeI2

Iron(II) iodide is used as a catalyst in organic reactions. It is also used as a compound in semiconductors and other high purity applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-86-0
Molecular Formula FeI2
MDL Number MFCD00016094
Synonym iron iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis

Specifications

Color Gray
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,diethyl ether and ethanol.
Formula Weight 309.66
Odor Odorless
Sensitivity Air and light sensitive; Hygroscopic
Appearance Gray beads
Density 5.32 g/cm3
Chemical Name or Material Iron(II) iodide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) iodide anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.