missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) iodide, anhydrous, 97% (metals basis)

Product Code. 11367147
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367147 10 g 10g
11357147 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367147 Supplier Thermo Scientific Alfa Aesar Supplier No. 011485.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-86-0

Iron(II) iodide is used as a catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-86-0
MDL Number MFCD00016094
Synonym iron iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis

Specifications

Color Brown
Physical Form Crystalline
Quantity 10 g
UN Number UN3260
Solubility Information Soluble in water,diethyl ether and ethanol.
Formula Weight 309.66
Percent Purity 97%
Odor Odorless
Sensitivity Air and light sensitive; Hygroscopic
Density 5.32 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.