missing translation for 'onlineSavingsMsg'
Learn More

Iron(II,III) oxide, nanopowder, 97%, Thermo Scientific Chemicals

Catalog Number 15452867
Click to view available options
Quantity:
25 g
100 g
500 g
1317-61-9
Fe3O4
231.53
MFCD00011010
WTFXARWRTYJXII-UHFFFAOYSA-N
diiron(3+) λ²-iron(2+) tetraoxidandiide
[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
1317-61-9
Fe3O4
231.53
MFCD00011010
WTFXARWRTYJXII-UHFFFAOYSA-N
diiron(3+) λ²-iron(2+) tetraoxidandiide
[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Iron(II,III) oxide is mainly used as a black-pigment-catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning and as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymers, rubber, fillers, building, construction, appliances and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1538°C
Color Black
Physical Form Powder
Quantity 500 g
Merck Index 14,4040
Solubility Information Insoluble in water and organic solvents. Soluble in concentrated mineral acids.
Formula Weight 231.54
Percent Purity 97%
Odor Odorless
Refractive Index 3.0
Density 5.2 g/mL
Chemical Name or Material Iron(II,III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'