missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) fluoride, anhydrous, 98%

Product Code. 11377147
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377147 10 g 10g
11387147 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377147 Supplier Thermo Scientific Alfa Aesar Supplier No. 011486.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-28-8 | F2Fe | 93.84 g/mol

Iron(II) fluoride is used for proteomics research applications. It is also a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-28-8
Molecular Formula F2Fe
Molecular Weight (g/mol) 93.84
MDL Number MFCD00016093
InChI Key FZGIHSNZYGFUGM-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) difluoride
SMILES [F-].[F-].[Fe++]

Specifications

Melting Point >1000°C (sublimation)
Color White
Physical Form Powder
Quantity 10 g
UN Number UN3260
Merck Index 14,4045
Solubility Information Insoluble in water.
Formula Weight 93.84
Percent Purity 98%
Sensitivity Air sensitive; Hygroscopic
Density 4.09 g/mL
Chemical Name or Material Iron(II) fluoride, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) fluoride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.