missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Chloride Tetrahydrate, TRC

Product Code. 30394540
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Product Code. Quantity
30394540 5 g
30393748 10 g
30396747 25 g
3 options
This item is not returnable. View return policy

Product Code. 30394540

Brand: Toronto Research Chemicals TRCI7749305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-10-9
Formula Weight 197.9149
InChI Formula InChI=1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2
IUPAC Name iron(2+);dichloride;tetrahydrate
Molecular Formula FeCl2H8O4
Molecular Weight (g/mol) 198.81
SMILES O.O.O.O.[Cl-].[Cl-].[Fe+2]
Synonym Ferrous Chloride Tetrahydrate, Iron Chloride Tetrahydrate, Iron Dichloride Tetrahydrate
Chemical Name or Material Iron(II) Chloride Tetrahydrate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.