missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Chloride Tetrahydrate, TRC

Product Code. 30393748
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393748 10 g
30394540 5 g
30396747 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393748 Supplier Toronto Research Chemicals Supplier No. TRCI77493010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-10-9
Molecular Formula FeCl2H8O4
Molecular Weight (g/mol) 198.81
Synonym Ferrous Chloride Tetrahydrate, Iron Chloride Tetrahydrate, Iron Dichloride Tetrahydrate
IUPAC Name iron(2+);dichloride;tetrahydrate
SMILES O.O.O.O.[Cl-].[Cl-].[Fe+2]

Specifications

Quantity 10 g
InChI Formula InChI=1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2
Formula Weight 197.9149
Chemical Name or Material Iron(II) Chloride Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.