missing translation for 'onlineSavingsMsg'
Learn More

Iron(Ii) Chloride Tetrahydrate, Honeywell™

Product Code. p-7071887 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15662070

Brand: Honeywell Research Chemicals 220299250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.805
MDL Number MFCD00149709
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl

Specifications

Quantity 250 g
Linear Formula FeCl2 · 4H2O
UN Number 3260
Formula Weight 198.81g/mol
Percent Purity 98%
Grade ReagentPlus™
Vapor Pressure 10mmHg (693°C)
Chemical Name or Material Iron(Ii) Chloride Tetrahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.