missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride hydrate, Reagent Grade, 99%

Product Code. 11371829
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
25g
5kg
This item is not returnable. View return policy

Product Code. 11371829

Brand: Thermo Scientific Alfa Aesar 012484.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Air Sensitive | CAS: 23838-02-0 | Cl2Fe . x H2O | 126.75 g/mol

It is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in wastewater treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23838-02-0
Molecular Formula Cl2Fe . x H2O
Molecular Weight (g/mol) 126.75
MDL Number MFCD00149711
InChI Key NMCUIPGRVMDVDB-UHFFFAOYSA-L
Synonym Iron dichloride, Iron protochloride
IUPAC Name λ²-iron(2+) dichloride
SMILES [Cl-].[Cl-].[Fe++]

Specifications

Color Green-Yellow
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99%
Solubility Information Soluble in water,alcohol and acetone. Insoluble in ether.
Formula Weight 126.75 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Iron(II) chloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.