missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride, anhydrous, 99.5% (metals basis)

Product Code. 11347418
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 11347418

Brand: Thermo Scientific Alfa Aesar 031141.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol

Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-94-3
Molecular Formula Cl2Fe
Molecular Weight (g/mol) 126.75
MDL Number MFCD00011004
InChI Key NMCUIPGRVMDVDB-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) dichloride
SMILES [Cl-].[Cl-].[Fe++]

Specifications

Boiling Point 1023°C
Melting Point 674°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Freely soluble in water,alcohol,acetone.Soluble in water,alcohol and acetone. Slightly soluble in benzene. Insoluble in ether.
Formula Weight 126.75
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Off-white powder
Density 3.16
Chemical Name or Material Iron(II) chloride, anhydrous
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.