missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11355787
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355787 100 g 100g
11365787 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355787 Supplier Thermo Scientific Alfa Aesar Supplier No. 043472.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as a Polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name iron(2+);dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 684°C
Physical Form Powder
Quantity 100 g
UN Number UN3260
Merck Index 14,4042
Solubility Information It is soluble in water and other solvents such as THF,methanol,ethanol.
Formula Weight 215.67
Percent Purity 99.995%
Odor Odorless
Packaging Ampule
Sensitivity Air sensitive; Hygroscopic
Density 4.636 g/mL
Assay (metals basis)
Chemical Name or Material Iron (II) bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.