missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, anhydrous, 98% min

Product Code. 11387418
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387418 100 g 100g
11367418 5 g 5g
11377418 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387418 Supplier Thermo Scientific Alfa Aesar Supplier No. 031154.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol

Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name λ²-iron(2+) dibromide
SMILES [Fe++].[Br-].[Br-]

Specifikationer

Melting Point 684°C
Color Yellow
Physical Form Granules
Quantity 100 g
Assay Percent Range 98% min.
Solubility Information Soluble in water,methanol,ethanol and tetrahydrofuran.
Formula Weight 215.67
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow granules
Density 4.636
Chemical Name or Material Iron(II) bromide, anhydrous
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.