Learn More
Iron(II) bromide, anhydrous, 98% min
Description
Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 684°C |
| Color | Yellow |
| Physical Form | Granules |
| Quantity | 25 g |
| Assay Percent Range | 98% min. |
| Solubility Information | Soluble in water,methanol,ethanol and tetrahydrofuran. |
| Formula Weight | 215.67 |
| Odor | Odorless |
| Sensitivity | Air sensitive; Hygroscopic |
| Appearance | Yellow granules |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.