missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, 98%, anhydrous

Product Code. 10320584 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10320584

Brand: Thermo Scientific 388720500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
SMILES [Fe++].[Br-].[Br-]

Specifications

Boiling Point 937°C
Melting Point 684°C
Color Black to Brown-Red
Physical Form Crystals or Powder
Quantity 50 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula FeBr2
Merck Index 15, 4069
Solubility Information Solubility in water: 1090g/L
IUPAC Name iron(2+);dibromide
Formula Weight 215.65
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Iron(II) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.