missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) acetate, anhydrous, Fe 29.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Iron(II) acetate is used as a mordant by the dye industry, as a catalyst in organic oxidation reactions and in carbon nanotube synthesis. It is used as a precursor in the production of other iron compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 190°C to 200°C (decomposition) |
Color | Brown |
Physical Form | Lump/Powder |
Quantity | 100 g |
Solubility Information | Soluble in water. |
IUPAC Name | λ²-iron(2+) diacetate |
Formula Weight | 173.94 |
Odor | Characteristic |
Sensitivity | Air and moisture sensitive |
Chemical Name or Material | Iron(II) acetate, anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'