missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane, Thermo Scientific™

Product Code. p-7051162
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 11308558

Brand: Thermo Scientific Alfa Aesar 040584.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331686-36-3
Molecular Formula C11H24FeO3
Molecular Weight (g/mol) 260.155
MDL Number MFCD00798541
InChI Key VRCGRDMZKVCBSY-UHFFFAOYSA-N
Synonym iron ii 2-ethylhexano-isopropoxide,in hexane, iron ii 2-ethylhexano-isopropoxide w/v in hexane, iron ii 2-ethylhexanoate monoisopropoxide w/v in hexane
PubChem CID 121233721
IUPAC Name 2-ethylhexanoic acid;iron;propan-2-ol
SMILES CCCCC(CC)C(=O)O.CC(C)O.[Fe]

Specifications

Quantity 25 mL
UN Number UN1208
Sensitivity Moisture sensitive
Formula Weight 258.14
Concentration or Composition (by Analyte or Components) 5% w/v in hexane
Physical Form Liquid
Chemical Name or Material Iron(II) 2-ethylhexanoate monoisopropoxide
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361f-Suspected of damaging fertility.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P315-Get immediate medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.