missing translation for 'onlineSavingsMsg'
Learn More

Iron disilicide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049331
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
12022-99-0
MFCD00016087
This item is not returnable. View return policy
12022-99-0
MFCD00016087

Iron disilicide is used for chemical research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1220°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 112.02
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Iron disilicide
compliance-icons
Product Identifier
  • Iron disilicide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'