missing translation for 'onlineSavingsMsg'
Learn More

Iron boride, 98%

Product Code. 11399199
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11399199

Brand: Thermo Scientific Alfa Aesar 088146.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. | CAS: 12006-84-7 | BFe | 66.66 g/mol

Iron boride is used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. Being inexpensive as well as having excellent mechanical, frictional and corrosion resistant properties, it can be used as replacement of ceramic or cermet type of materials in fabrication and machining. It is also involved in the production of borated steels and borated stainless steels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-84-7
Molecular Formula BFe
Molecular Weight (g/mol) 66.66
MDL Number MFCD00016092
InChI Key UJYBTTFGLAOKMN-UHFFFAOYSA-N
Synonym iron boride, iron boride feb, ferrous boride, ironylidyneborane, ferrioborane, ferric boride
PubChem CID 82789
SMILES [B+]=[Fe-]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name ferrioborane
Formula Weight 66.66
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 7.15
Density 7.15
Chemical Name or Material Iron boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.