missing translation for 'onlineSavingsMsg'
Learn More

Iron arsenide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11343228
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343228 10 g 10g
11333228 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11343228 Supplier Thermo Scientific Alfa Aesar Supplier No. 036190.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Iron arsenide is used as a superconductor, semiconductor and in photo optic applications 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-88-8
Molecular Formula AsFe2
Molecular Weight (g/mol) 186.612
MDL Number MFCD00151533
InChI Key FDKAYGUKROYPRO-UHFFFAOYSA-N
Synonym 1-arsa-2,3-diferracycloprop-1-ene, iron arsenide, pieces trace metals basis
PubChem CID 71310806
SMILES [Fe].[Fe]=[As]

Specifications

Physical Form Lump
Quantity 10 g
UN Number UN1557
Solubility Information It is insoluble in water.
Formula Weight 186.62
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Iron arsenide
compliance-icons compliance-icons
Product Identifier
  • Iron arsenide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.