missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron for AAS 1g/L, 12% ethanol matrix, Fisher chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
Specifications
Specifications
| Packaging | Bottle |
| Physical Form | Liquid |
| Concentration | 1 g/L±0.002 g/L |
| UN Number | 1993 |
| Type | Flame Single-element Standard Solution |
| Solvent or Matrix | 12% Ethanol |
| Quantity | 100 mL |
| Chemical Name or Material | Iron |
Safety and Handling
Product Identifier
- Single-element Standard Solution for AAS. Iron (Fe) 1000mg/l in Ethanol 12%,HCL 2%
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction