missing translation for 'onlineSavingsMsg'
Learn More

Iron(0) pentacarbonyl

Product Code. 10695491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10695491 25 g Glass bottle 25g
10278333 100 g Glass bottle 100g
10175323 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10695491 Supplier Thermo Scientific Acros Supplier No. 412600250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 13463-40-6 | C5FeO5 | 195.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-40-6
Molecular Formula C5FeO5
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011001
InChI Key FYOFOKCECDGJBF-UHFFFAOYSA-N
Synonym Pentacarbonyl iron
IUPAC Name pentakis(methanidylidyneoxidanium) iron
SMILES [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Specifications

Boiling Point 103.0°C
Melting Point -20.0°C
Color Brown to Orange-Yellow
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99.50%
Linear Formula Fe(CO)5
Fieser 01,519; 02,229; 03,167; 04,268; 05,357; 06,304; 10,221; 13,152; 17,270
Flash Point −15°C
Merck Index 15, 5143
Solubility Information Solubility in water: insoluble
Formula Weight 195.9
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.49
Density 1.4900g/mL
Chemical Name or Material Iron(0) pentacarbonyl, 99.5%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iron pentacarbonyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.