missing translation for 'onlineSavingsMsg'
Learn More

Irisolidone, TRC

Product Code. 30390126
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390126 2.5 mg 2.5mg
30408160 1 mg -
30388947 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390126 Supplier Toronto Research Chemicals Supplier No. TRCI2484752.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2345-17-7
Molecular Formula C17H14O6
Molecular Weight (g/mol) 314.29
IUPAC Name 5,7-dihydroxy-6-methoxy-3-(4-methoxyphenyl)chromen-4-one
SMILES COc1ccc(cc1)C2=COc3cc(O)c(OC)c(O)c3C2=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H14O6/c1-21-10-5-3-9(4-6-10)11-8-23-13-7-12(18)17(22-2)16(20)14(13)15(11)19/h3-8,18,20H,1-2H3
Formula Weight 314.079
Chemical Name or Material Irisolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.