missing translation for 'onlineSavingsMsg'
Learn More

Irish Moss Carrageenan, MP Biomedicals™

Product Code. 11421762 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421762 1 kg 1kg
11401762 100 g 100g
11411762 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421762 Supplier MP Biomedicals Supplier No. 0210488691
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irish Moss Carrageenan solution is used to suppress immune response in vivo and in vitro via mechanisms.

  • Carrageenans are used to suppress immune response in vivo and in vitro via mechanisms believed to involve selective cytopathic effect on macrophages.
  • As ι-, κ-, and λ-carrageenans have varying degress of sulfation, they may be studied comparatively for differences in protective, antiviral, and immunosuppression activities.
  • Carrageenan may be used in the development of biomaterials such as bone graft substitutes, for entrapping vessels such as biobeads, and as encapsulation vehicles for drug delivery.
  • Carrageenan can be used to develop hydrogels.

Chemical Identifiers

CAS 9000-07-1
MDL Number MFCD00081480
Synonym Carrageenan, Gelatin

Specifications

Color Tan
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Irish Moss (Carrageenan), Predominantly Kappa-Carrageenan with lesser amount Lambda-Carrageenan.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.