missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride trihydrate, MedChemExpress

Product Code. 16427126
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16427126

Brand: Medchem Express HY16568

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Irinotecan hydrochloride trihydrate ((+)-Irinotecan hydrochloride trihydrate) is a topoisomerase I inhibitor with antitumor activity.

Irinotecan hydrochloride trihydrate ((+)-Irinotecan hydrochloride trihydrate) is a topoisomerase I inhibitor with antitumor activity.

Specifications

Chemical Name or Material Irinotecan hydrochloride trihydrate
CAS 136572-09-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C33H45ClN4O9
Quantity 100 mg
Synonym (+)-Irinotecan hydrochloride trihydrateCPT-11 hydrochloride trihydrate
Solubility Information DMSO : 50 mg/mL (73.84 mM; Need ultrasonic) ∣H2O : 1.52 mg/mL (2.24 mM; Need ultrasonic and warming)
SMILES [H]Cl.O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3.O.O.O
Molecular Weight (g/mol) 677.18
Formula Weight 677.18
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.