missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan Carboxylate Sodium Salt, TRC

Product Code. 30391239
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391239 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391239 Supplier Toronto Research Chemicals Supplier No. TRCI76751050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Irinotecan Carboxylate Sodium Salt
CAS 1329502-92-2
Molecular Formula C33H39N4O7.Na
Quantity 50 mg
Synonym Indolizino[1,2-b]quinoline-7-acetic acid, 2-[([1,4'-bipiperidin]-1'-ylcarbonyl)oxy]-α,12-diethyl-9,11-dihydro-α-hydroxy-8-(hydroxymethyl)-9-oxo-, sodium salt (1:1), (αS)-, Irinotecan Carboxylate Sodium Salt, (alphaS)-2-[([1,4'-Bipiperidin]-1'-ylcarbonyl)oxy]-alpha,12-diethyl-9,11-dihydro-alpha-hydroxy-8-(hydroxymethyl)-9-oxo-indolizino[1,2-b]quinoline-7-acetic Acid Sodium Salt, CPT 11 Carboxylic Acid Sodium Salt
InChI Formula InChI=1S/C33H40N4O7.Na/c1-3-22-23-16-21(44-32(42)36-14-10-20(11-15-36)35-12-6-5-7-13-35)8-9-27(23)34-29-24(22)18-37-28(29)17-26(25(19-38)30(37)39)33(43,4-2)31(40)41;/h8-9,16-17,20,38,43H,3-7,10-15,18-19H2,1-2H3,(H,40,41);/q;+1/p-1/t33-;/m0./s1
SMILES [Na+].CCc1c2CN3C(=O)C(=C(C=C3c2nc4ccc(OC(=O)N5CCC(CC5)N6CCCCC6)cc14)[C@@](O)(CC)C(=O)[O-])CO
IUPAC Name sodium;(2S)-2-[12-ethyl-8-(hydroxymethyl)-9-oxo-2-(4-piperidin-1-ylpiperidine-1-carbonyl)oxy-11H-indolizino[1,2-b]quinolin-7-yl]-2-hydroxybutanoate
Molecular Weight (g/mol) 626.68
Formula Weight 626.2716
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.