missing translation for 'onlineSavingsMsg'
Learn More

Iridium wire, 0.075mm (0.003in) dia, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11469940
Click to view available options
Quantity:
10 cm
50 cm
This item is not returnable. View return policy
7439-88-5
Ir
192.22
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
23924
CHEBI:49666
iridium
[Ir]
This item is not returnable. View return policy
7439-88-5
Ir
192.22
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
23924
CHEBI:49666
iridium
[Ir]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 cm
Percent Purity 99.9%
Weight ≈0.099g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Iridium wire, 0.075mm (0.003 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'