missing translation for 'onlineSavingsMsg'
Learn More

Iridium sponge, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11396418
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
7439-88-5
Ir
192.22
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
23924
CHEBI:49666
iridium
[Ir]
This item is not returnable. View return policy
7439-88-5
Ir
192.22
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
23924
CHEBI:49666
iridium
[Ir]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3089
Quantity 2 g
Percent Purity 99.95%
Mesh Size −100 Mesh
Form Sponge
Assay (metals basis)
Chemical Name or Material Iridium sponge

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'