missing translation for 'onlineSavingsMsg'
Learn More

Iridium, plasma standard solution, Specpure™ Ir 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7050166
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00011062
This item is not returnable. View return policy
MFCD00011062

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration IrCl3 in 20% HCl
Quantity 50 mL
UN Number UN1789
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Iridium plasma standard solution, Ir 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'