missing translation for 'onlineSavingsMsg'
Learn More

Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL, Thermo Scientific Chemicals

Product Code. 11318637
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Quantity 50 mL
Odor Characteristic
Physical Form Liquid
Chemical Name or Material Iridium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'