Learn More
Iridium(IV) oxide dihydrate, Premion™, 99.99% (metals basis), Ir 73% min, Thermo Scientific Chemicals
£520.00 - £2290.00
Chemical Identifiers
CAS | 30980-84-8 |
---|---|
Molecular Formula | H2IrO3 |
Molecular Weight (g/mol) | 242.23 |
MDL Number | MFCD00011065 |
InChI Key | YEPXCFHWKVNVAX-UHFFFAOYSA-N |
Synonym | iridium iv oxide hydrate, iro2.h2o, dioxoiridium hydrate, dioxoiridium-water 1/1, iridium iv oxide hydrate trace metals basis |
PubChem CID | 16212540 |
IUPAC Name | dioxoiridium;hydrate |
SMILES | O.O=[Ir]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314708
|
Thermo Scientific Alfa Aesar
043051.03 |
1 g |
£520.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304708
|
Thermo Scientific Alfa Aesar
043051.06 |
5 g |
£2290.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iridium(IV) oxide dihydrate is used with other rare oxides in the coating of anode-electrodes for industrial electrolysis and in microelectrodes for electrophysiology research. It is also used for reference electrode, which finds application in various fields biology, food industry, nuclear field, oil and gas industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30980-84-8 | |
242.23 | |
YEPXCFHWKVNVAX-UHFFFAOYSA-N | |
16212540 | |
O.O=[Ir]=O |
H2IrO3 | |
MFCD00011065 | |
iridium iv oxide hydrate, iro2.h2o, dioxoiridium hydrate, dioxoiridium-water 1/1, iridium iv oxide hydrate trace metals basis | |
dioxoiridium;hydrate |
Specifications
30980-84-8 | |
H2IrO3 | |
iridium iv oxide hydrate, iro2.h2o, dioxoiridium hydrate, dioxoiridium-water 1/1, iridium iv oxide hydrate trace metals basis | |
YEPXCFHWKVNVAX-UHFFFAOYSA-N | |
dioxoiridium;hydrate | |
16212540 | |
99.99% | |
Ir 73% min | |
Iridium (IV) oxide dihydrate |
Powder | |
MFCD00011065 | |
Insoluble in water. | |
O.O=[Ir]=O | |
242.23 | |
260.23 (224.20 Anhydrous) | |
Premion™ | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 234-743-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only