Learn More
Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5 min
Description
Iridium(IV) chloride is used in chemical analysis as testing for nitric acid in the presence of nitrous acid, microscopy and in plating solutions. It acts as a catalyst precursor for the stereoselective reduction of cyclohexanones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Physical Form | Glassy Amorphous Pieces, Powder |
| Quantity | 1 g |
| Assay Percent Range | 99.95% (metals basis) |
| Solubility Information | Soluble in water,alcohol and dilute hydrochloric acid. |
| Formula Weight | 334.01 |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Iridium(IV) chloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.