missing translation for 'onlineSavingsMsg'
Learn More

Iridium(IV) bromide, Premion™, 99.99% (metals basis), Ir 37.1% min

Product Code. p-7050800
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy

Product Code. 11398288

Brand: Thermo Scientific Alfa Aesar 039493.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-64-2 | Br4Ir | 511.833 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-64-2
Molecular Formula Br4Ir
Molecular Weight (g/mol) 511.833
MDL Number MFCD00049467
InChI Key IYWTUWKWQJIZPO-UHFFFAOYSA-J
Synonym iridium bromide irbr4, iridium iv bromide, premion, iridium iv bromide,premion metalsbasis ,irmin
PubChem CID 14029372
IUPAC Name tetrabromoiridium
SMILES Br[Ir](Br)(Br)Br

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 0.5 g
Solubility Information Soluble in water
Formula Weight 511.84
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Ir 37.1% min.
Odor Acrid
Assay (metals basis)
Chemical Name or Material Iridium(IV) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.