missing translation for 'onlineSavingsMsg'
Learn More

Iridium(IV) bromide, Premion™, 99.99% (metals basis), Ir 37.1% min

Product Code. 11398288
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398288 0.5 g 0.5g
11308298 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398288 Supplier Thermo Scientific Alfa Aesar Supplier No. 039493.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-64-2 | Br4Ir | 511.833 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-64-2
Molecular Formula Br4Ir
Molecular Weight (g/mol) 511.833
MDL Number MFCD00049467
InChI Key IYWTUWKWQJIZPO-UHFFFAOYSA-J
Synonym iridium bromide irbr4, iridium iv bromide, premion, iridium iv bromide,premion metalsbasis ,irmin
PubChem CID 14029372
IUPAC Name tetrabromoiridium
SMILES Br[Ir](Br)(Br)Br

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 0.5 g
Solubility Information Soluble in water
Formula Weight 511.84
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Ir 37.1% min.
Odor Acrid
Assay (metals basis)
Chemical Name or Material Iridium(IV) bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.