missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) chloride hydrate, 99.8% (metals basis)

Product Code. 11371648
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11371648

Brand: Thermo Scientific Alfa Aesar 011030.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14996-61-3 | Cl3Ir | 298.57 g/mol

Iridium(III) chloride hydrate is used to prepare other iridium compounds. It finds use as a catalyst for oxidation in presence of cerium(IV) and in the the manufacture of electrodes. Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of Paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14996-61-3
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.57
MDL Number MFCD00149750
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
IUPAC Name iridium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ir+3]

Specifications

Color Black
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water and alcohol.
Formula Weight 298.56 (Anhydrous)
Odor Odorless
Chemical Name or Material Iridium(III) chloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.