missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) chloride, anhydrous, Ir 62% min

Product Code. 11356528
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356528 5 g 5g
11346528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356528 Supplier Thermo Scientific Alfa Aesar Supplier No. 012158.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-83-9 | Cl3Ir | 298.57 g/mol

Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-83-9
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.57
MDL Number MFCD00011063
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
Synonym iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, acmc-1cp8w, iridium iv chloride, premion
PubChem CID 24815
IUPAC Name tetrachloroiridium
SMILES [Cl-].[Cl-].[Cl-].[Ir+3]

Specifications

Melting Point 763°C (decomposition)
Color Black to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.95% (Metal basis)
Solubility Information It is insoluble in water.
Formula Weight 298.56
Sensitivity Hygroscopic
Appearance Drak brown to black glassy amorphous pieces/powder
Chemical Name or Material Iridium(IV) chloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.