missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) bromide hydrate, Ir 35.9% min

Product Code. 11356618
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356618 1 g 1g
11366618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356618 Supplier Thermo Scientific Alfa Aesar Supplier No. 012212.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 171769-74-7 | Br3Ir | 431.93 g/mol

Iridium(III) bromide hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171769-74-7
Molecular Formula Br3Ir
Molecular Weight (g/mol) 431.93
MDL Number MFCD00049466
InChI Key HTFVQFACYFEXPR-UHFFFAOYSA-K
SMILES [Br-].[Br-].[Br-].[Ir+3]

Specifications

Melting Point 100°C (in -3H2O)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name iridium(3+) tribromide
Formula Weight 431.91 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ir 35.9% min.
Chemical Name or Material Iridium(III) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.