missing translation for 'onlineSavingsMsg'
Learn More

Iridium foil, 0.5mm (0.02in) thick, typically 99.7+% (metals basis), Thermo Scientific Chemicals

Catalog Number 11343218
Click to view available options
Quantity:
25 x 25 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7439-88-5
Molecular Formula Ir
MDL Number MFCD00011062
Quantity 25 x 25 mm
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES [Ir]
IUPAC Name iridium
Molecular Weight (g/mol) 192.22
PubChem CID 23924
ChEBI CHEBI:49666
Percent Purity ≥99.7% (typically)
Weight ≈7g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Iridium foil, 0.5mm (0.02 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'