missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRGM Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76377
This item is not returnable.
View return policy
Description
IRGM Polyclonal specifically detects IRGM in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| IRGM | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| IFI1, immunity-related GTPase family M protein, immunity-related GTPase family, M, interferon-inducible protein 1, IRGM1, LPS-stimulated RAW 264.7 macrophage protein 47 homolog, LRG47, LRG-47, LRG-47-like protein, M1, MGC149263, MGC149264 | |
| Rabbit | |
| 20 kDa | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
| AAI28169 | |
| IRGM | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of the human IRGM. The immunogen is located within amino acids 30 - 80 of IRGM. Amino Acid Squence: ISALRNGTHEGKASPPTE | |
| Affinity Purified | |
| RUO | |
| 345611 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction