missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress iRGD peptide

Product Code. 16411828
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411828 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411828 Supplier MedChemExpress Supplier No. HYP0122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress iRGD peptide is a 9-amino acid cyclic peptide, triggers tissue penetration of drugs by first binding to av integrins, then proteolytically cleaved in the tumor to produce CRGDK/R to interact with neuropilin-1, and has tumor-targeting and tumor-penetrating properties.

Specifications

Purity 98.79%
CAS 1392278-76-0
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 948.04 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : ≥ 50 mg/mL (52.74 mM)
Synonym c(CRGDKGPDC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.