missing translation for 'onlineSavingsMsg'
Learn More

IRGC Antibody, DyLight 755, Novus Biologicals™

Product Code. 18400675 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400675 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400675 Supplier Novus Biologicals Supplier No. NBP177364IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRGC Polyclonal antibody specifically detects IRGC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRGC
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CINEMA, EC 3.6.5, EC 3.6.5.-, Iigp5, Immunity-related GTPase cinema 1, immunity-related GTPase family, cinema, immunity-related GTPase family, cinema 1, interferon inducible GTPase 5, interferon-inducible GTPase 5, IRGC1, R30953_1
Host Species Rabbit
Immunogen Antibody was raised against a 12 amino acid synthetic peptide near the carboxy terminus of human IRGC. The immunogen is located within the last 50 amino acids of IRGC. Amino Acid Squence: CLDSWKKHDSEEK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 56269
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.