missing translation for 'onlineSavingsMsg'
Learn More

Irganox 1310, Dr. Ehrenstorfer

Product Code. 30328185
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328185 Supplier Dr. Ehrenstorfer Supplier No. DREC14373970
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylpropionic Acid
CAS 20170-32-5
Analyte or Component Names 3,5-Di-tert-butyl-4-hydroxyphenylpropionic Acid
Shipping Condition +20°C
Molecular Formula C17H26O3
Certifications/Compliance ISO/IEC 17025
Synonym 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic Acid, 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoic Acid, 3,5-Di-tert-butyl-4-hydroxybenzenepropanoic Acid, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic Acid, 3,5-Di-tert-butyl-4-hydroxyphenylpropionic Acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propanoic Acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid, 3-(3',5'-Di-tert-butyl-4'-hydroxyphenyl)propionic Acid, 4-Hydroxy-3,5-di-tert-butylphenylpropionic Acid, Fenoksan, Fenoxan, Fenozan Fenozan Acid, Irganox 1310, Irganox B 2, Phenosan, Phenoxan, Phenoxan (antioxidant), Phenozan, β-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid β-(4-Hydroxy-3,5-di-tert-butylphenyl)propanoic Acid, β-(4-Hydroxy-3,5-di-tert-butylphenyl)propionic Acid
SMILES CC(C)(C(C=C(CCC(O)=O)C=C1C(C)(C)C)=C1O)C
Molecular Weight (g/mol) 278.39
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.