missing translation for 'onlineSavingsMsg'
Learn More

Irganox 1035, TRC

Product Code. 30408253
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408253 100 g 100g
30391238 50 g 50g
30388783 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408253 Supplier Toronto Research Chemicals Supplier No. TRCI764153100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41484-35-9
Molecular Formula C38 H58 O6 S
Molecular Weight (g/mol) 642.93
Synonym Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,1'-(thiodi-2,1-ethanediyl) ester, Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, thiodi-2,1-ethanediyl ester (9CI), 2,2-Thiodiethylene bis-[(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 2,2'-Thiodiethyl bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 2,2'-Thiodiethylene bis[3-(3,5-di-tert-butyl-4-hydroxyhydrocinnamate], 2,2'-Thiodiethylene bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 2-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 2,2'-thiobisethanol diester, 3,5-Di-tert-butyl-4-hydroxybenzenepropanoic acid thiodi-2,1-ethanediyl ester, 3-Thia-1,5-pentanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], AO 75, Anox 70, Antioxidant 1035, Bis[(3,5-di-tert-butyl-4-hydroxyphenyl)propionyl-2-oxyethyl] sulfide, Bis[2-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]ethyl] sulfide, Chinox 1035, Chinox S 35, Durad AX 15, Fenozan 30, IR 1035, Irganox 1035, Irganox 1035FF, Irganox L 1035, Irganox L 115, L 115, Naugard EL 50, S 35, Thiodi(2,1-ethanediyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Thiodiethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Thiodiethylene bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Thiodiethylene glycol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Thiodiethylene glycol bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Thiodiethylene glycol bis[β-(4-hydroxy-3,5-di(tert-butyl)phenyl)propionate]
IUPAC Name 2-[2-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxy]ethylsulfanyl]ethyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
SMILES CC(C)(C)c1cc(CCC(=O)OCCSCCOC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C38H58O6S/c1-35(2,3)27-21-25(22-28(33(27)41)36(4,5)6)13-15-31(39)43-17-19-45-20-18-44-32(40)16-14-26-23-29(37(7,8)9)34(42)30(24-26)38(10,11)12/h21-24,41-42H,13-20H2,1-12H3
Formula Weight 642.3954
Chemical Name or Material Irganox 1035
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.