missing translation for 'onlineSavingsMsg'
Learn More

Irgacure 127, TRC

Product Code. 30394779
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394779 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394779 Supplier Toronto Research Chemicals Supplier No. TRCI764155500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis[4-(2-hydroxy-2-methylpropionyl)phenyl]methane
CAS 474510-57-1
Molecular Formula C21 H24 O4
Quantity 500 mg
Synonym 1,1'-(Methylene-di-4,1-phenylene)bis[2-hydroxy-2-methyl-1-propanone], 2-Hydroxy-1-[4-[4-(2-hydroxy-2-methylpropionyl)benzyl]phenyl]-2-methylpropan-1-one, Bis[4-(2-hydroxy-2-methylpropionyl)phenyl]methane, Irgacure 127
InChI Formula InChI=1S/C21H24O4/c1-20(2,24)18(22)16-9-5-14(6-10-16)13-15-7-11-17(12-8-15)19(23)21(3,4)25/h5-12,24-25H,13H2,1-4H3
SMILES CC(C)(O)C(=O)c1ccc(Cc2ccc(cc2)C(=O)C(C)(C)O)cc1
IUPAC Name 2-hydroxy-1-[4-[[4-(2-hydroxy-2-methylpropanoyl)phenyl]methyl]phenyl]-2-methylpropan-1-one
Molecular Weight (g/mol) 340.413
Formula Weight 340.167
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.