missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF9 Monoclonal Antibody (K70047_18H9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556264
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: REDQDAAFFK AWAIFKGKYK EGDTGGPAVW KTRLRCALNK SSEFKEVPER GRMDVAEPYK VYQLLPPGIV SGQPGTQKVP SKRQHSSVSS ERKEEEDAMQ NCTLSPSVLQ DSLNNEEEGA SGGAVHSDIG SSSSSSSPEP QEVTDTTEAP FQGDQRSLEF
Transcription factor that plays an essential role in anti-viral immunity. It mediates signaling by type I IFNs (IFN-alpha and IFN-beta). Following type I IFN binding to cell surface receptors, Jak kinases (TYK2 and JAK1) are activated, leading to tyrosine phosphorylation of STAT1 and STAT2. IRF9/ISGF3G associates with the phosphorylated STAT1:STAT2 dimer to form a complex termed ISGF3 transcription factor, that enters the nucleus. ISGF3 binds to the IFN stimulated response element (ISRE) to activate the transcription of interferon stimulated genes, which drive the cell in an antiviral state. [UniProt].
Specifications
| IRF9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q00978 | |
| IRF9 | |
| Recombinant human IRF9 (immunogen range is 51-210). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K70047_18H9 | |
| Unconjugated | |
| IRF9 | |
| gamma 48kDa; IFN alpha responsive transcription factor subunit; IFN-alpha-responsive transcription factor subunit; interferon dependent positive acting transcription factor 3 gamma; interferon regulatory factor 9; Interferon-stimulated gene factor 3 gamma; interferon-stimulated transcription factor 3; interferon-stimulated transcription factor 3, gamma (48kD); interferon-stimulated transcription factor 3, gamma 48kDa; IRF9; Irf-9; ISGF3; ISGF-3 gamma; ISGF3 p48 subunit; ISGF3G; p48; transcriptional regulator ISGF3 subunit gamma; Unknown (protein for MGC:126912); Unknown (protein for MGC:92097); zgc:92097 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction