missing translation for 'onlineSavingsMsg'
Learn More

IRF9 Antibody (PCRP-IRF9-2F8), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18369405 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369405 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369405 Supplier Novus Biologicals Supplier No. NBP314228AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF9
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-IRF9-2F8
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma
Host Species Mouse
Immunogen Recombinant full-length human IRF9 protein (Uniprot: Q00978)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10379
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.