missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRF8 Recombinant Rabbit Monoclonal Antibody (K01_1T06)

Product Code. 30826382
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30826382 100 μL -
30825808 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826382 Supplier Invitrogen™ Supplier No. MA563860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

IRF8 Recombinant Monoclonal Antibody for Western Blot

IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF8
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Irf8
Gene Accession No. Q02556
Gene Alias AI893568; H-ICSBP; Icsbp; Icsbp1; IMD32A; IMD32B; interferon consensus sequence binding protein 1; Interferon consensus sequence-binding protein; Interferon regulatory factor 8; Irf8; IRF-8; Myls; transcription factor ICSBP
Gene Symbols Irf8
Host Species Rabbit
Immunogen Recombinant protein of human IRF8.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3394
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.