missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585224
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: THP-1 nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.
Specifications
| IRF8 | |
| Polyclonal | |
| Unconjugated | |
| Irf8 | |
| AI893568; H-ICSBP; Icsbp; Icsbp1; IMD32A; IMD32B; interferon consensus sequence binding protein 1; Interferon consensus sequence-binding protein; Interferon regulatory factor 8; Irf8; IRF-8; Myls; transcription factor ICSBP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q02556 | |
| Irf8 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human IRF8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction