Learn More
Invitrogen™ IRF8 Monoclonal Antibody (V3GYWCH), eFluor™ 450, eBioscience™

Mouse Monoclonal Antibody
£367.00
Specifications
| Antigen | IRF8 |
|---|---|
| Clone | V3GYWCH |
| Concentration | 0.2 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: The V3GYWCH monoclonal antibody reacts with human and mouse interferon regulatory factor 8 (IRF8; ICSBP). IRF8 is a 50 kDa transcription factor that plays a critical role in the development of dendritic cells (DC). Specifically, IRF8 is essential in the development and differentiation of plasmacytoid DC (pDC) and CD8a+ DC. IRF8-/- mice are deficient in both pDC and CD8a+ DC populations. CD8a- DC are present in normal numbers in IRF8-/- mice but fail to mature upon Toll-like receptor signaling and are functionally impaired in response to in vivo microbial stimulation. IRF8 deficiency does not affect the frequency or viability of DC precursors and as a result retroviral IRF8 transduction restores pDC development from DC progenitors in IRF8-/- mice. Applications Reported: This V3GYWCH antibody has been reported for use in flow cytometric analysis. Applications Tested: This V3GYWCH antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405...
IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.Specifications
| IRF8 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| eFluor 450 | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P23611, Q02556 | |
| 15900, 3394 | |
| Primary | |
| Affinity chromatography |
| V3GYWCH | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human, Mouse | |
| Irf8 | |
| AI893568; H-ICSBP; Icsbp; Icsbp1; IMD32A; IMD32B; interferon consensus sequence binding protein 1; Interferon consensus sequence-binding protein; Interferon regulatory factor 8; Irf8; IRF-8; Myls; transcription factor ICSBP | |
| Irf8 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.