Learn More
Invitrogen™ IRF8 Monoclonal Antibody (V3GYWCH), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£121.00 - £293.00
Specifications
| Antigen | IRF8 |
|---|---|
| Clone | V3GYWCH |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15528266
|
Invitrogen™
17-9852-80 |
25 μg |
£121.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15538266
![]() |
Invitrogen™
17-9852-82 |
100 μg |
£293.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The V3GYWCH monoclonal antibody reacts with human and mouse interferon regulatory factor 8 (IRF8; ICSBP). IRF8 is a 50 kDa transcription factor that plays a critical role in the development of dendritic cells (DC). Specifically, IRF8 is essential in the development and differentiation of plasmacytoid DC (pDC) and CD8a+ DC. IRF8-/- mice are deficient in both pDC and CD8a+ DC populations. CD8a- DC are present in normal numbers in IRF8-/- mice but fail to mature upon Toll-like receptor signaling and are functionally impaired in response to in vivo microbial stimulation. IRF8 deficiency does not affect the frequency or viability of DC precursors and as a result retroviral IRF8 transduction restores pDC development from DC progenitors in IRF8-/- mice. Applications Reported: This V3GYWCH antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This V3GYWCH antibody has been tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One-step protocol: intracellular (nuclear) proteins located under the Resources Tab online. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
IRF8 Monoclonal antibody specifically detects IRF8 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| IRF8 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AI893568; H-ICSBP; Icsbp; Icsbp1; IMD32A; IMD32B; interferon consensus sequence binding protein 1; Interferon consensus sequence-binding protein; Interferon regulatory factor 8; Irf8; IRF-8; Myls; transcription factor ICSBP | |
| Irf8 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Irf8 |
| V3GYWCH | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human, Mouse | |
| P23611, Q02556 | |
| 15900, 3394 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.