missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF7 Antibody - BSA Free, Novus Biologicals™
Description
IRF7 Polyclonal antibody specifically detects IRF7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
Specifications
| Antigen | IRF7 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 to 1 μg/mL, ELISA, Immunohistochemistry 2 to 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 to 5 μg/mL, Knockout Validated |
| Formulation | PBS |
| Gene Alias | interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a peptide corresponding to 17 amino acids near the center of human IRF7. The immunogen is located within amino acids 200-230 of IRF7. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?