missing translation for 'onlineSavingsMsg'
Learn More

IRF7 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18372382 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372382 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372382 Supplier Novus Biologicals Supplier No. NBP306987AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRF7 Polyclonal antibody specifically detects IRF7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF7
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the center of human IRF7. The immunogen is located within amino acids 200-230 of IRF7.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3665
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.